Methyl-6-indolcarboxylat ≥98%
Lieferant: Thermo Scientific
Synonyme:
Methyl 1H-indole-6-carboxylate
L16020.03
L16020.06
L16020.03EA
19
EUR
L16020.03
L16020.06
Methyl-6-indolcarboxylat ≥98%
Methyl-6-indolcarboxylat
Purity: 98%
CAS No.: 50820-65-0
Molecular Formula: C10H9NO2
Formula Weight: 175.19
Melting Point: 78-80°
Beilstein Reference: 141837
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,19 g/mol Schmelzpunkt: 78…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00211063 CAS-Nummer: 50820-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...