Methyl-6-indolcarboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 1H-indole-6-carboxylate
IN-0015-25G
IN-0015-5G
IN-0015-1G
COBBIN-0015-25GEA
127
EUR
COBBIN-0015-25G
COBBIN-0015-5G
COBBIN-0015-1G
Methyl-6-indolcarboxylat
Methyl-6-indolcarboxylat
Formel:
C₁₀H₉NO₂ Molecular Weight: 175,19 g/mol |
MDL:
MFCD00211063 CAS-Nummer: 50820-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...