1-Boc-5-aminoindol

Lieferant: CHEMPUR

Synonyme: 5-Amino-1H-indole, N1-BOC protected, 1-Boc-5-Aminoindole

48-0049.1 48-0049.5
CHMP48-0049.1EA 0 EUR
CHMP48-0049.1 CHMP48-0049.5
1-Boc-5-aminoindol
1-Boc-5-aminoindol
Formel: C₁₃H₁₆N₂O₂
Molecular Weight: 232.28 g/mol
MDL: MFCD04114746
CAS-Nummer: 166104-20-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR