1-Boc-6-aminoindol

Lieferant: CHEMPUR

Synonyme: tert-Butyl 6-amino-1H-indole-1-carboxylate

48-0048.1 48-0048.5
CHMP48-0048.1EA 0 EUR
CHMP48-0048.1 CHMP48-0048.5
1-Boc-6-aminoindol
1-Boc-6-aminoindol
Formel: C₁₃H₁₆N₂O₂
Molecular Weight: 232,28 g/mol
MDL: MFCD04114745
CAS-Nummer: 219508-62-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR