1-Boc-5-methoxyindol

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 5-methoxy-1H-indole-1-carboxylate, N-Boc-5-methoxyindole

IN-0140-5G IN-0140-25G IN-0140-1G
COBBIN-0140-5GEA 227 EUR
COBBIN-0140-5G COBBIN-0140-25G COBBIN-0140-1G
1-Boc-5-methoxyindol
1-Boc-5-methoxyindol
Formel: C₁₄H₁₇NO₃
Molecular Weight: 247.29 g/mol
MDL: MFCD08272248
CAS-Nummer: 99275-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR