1-Boc-5-methoxyindol, Sigma-Aldrich®

Synonyme: tert-Butyl 5-methoxy-1H-indole-1-carboxylate, N-Boc-5-methoxyindole

M-Clarity™ Qualitätsstufe = MQ100

680613-10G 680613-1G
SIAL680613-10GEA 368 EUR
SIAL680613-10G SIAL680613-1G
1-Boc-5-methoxyindol, Sigma-Aldrich®
1-Boc-5-methoxyindol
Formel: C₁₄H₁₇NO₃
Molecular Weight: 247.29 g/mol
MDL: MFCD08272248
CAS-Nummer: 99275-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR