1-Boc-7-methoxyindol

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 7-methoxy-1H-indole-1-carboxylate

IN-0223-5G IN-0223-1G
COBBIN-0223-5GEA 451 EUR
COBBIN-0223-5G COBBIN-0223-1G
1-Boc-7-methoxyindol
1-Boc-7-methoxyindol
Formel: C₁₄H₁₇NO₃
Molecular Weight: 247,29 g/mol
MDL: MFCD15143582
CAS-Nummer: 1215205-77-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR