1-Boc-5-iodindol
Lieferant: COMBI-BLOCKS
Synonyme:
5-Iodo-1H-indole, N-BOC protected
IN-0162-1G
IN-0162-5G
COBBIN-0162-1GEA
166
EUR
COBBIN-0162-1G
COBBIN-0162-5G
1-Boc-5-iodindol
1-Boc-5-iodindol
Formel:
C₁₃H₁₄INO₂ Molecular Weight: 343,16 g/mol |
MDL:
MFCD09258644 CAS-Nummer: 374818-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...