1-Boc-7-methylindol

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 7-methyl-1H-indole-1-carboxylate, 7-Methyl-1H-indole, N-BOC protected

IN-0155-5G IN-0155-1G
COBBIN-0155-5GEA 0 EUR
COBBIN-0155-5G COBBIN-0155-1G
1-Boc-7-methylindol
1-Boc-7-methylindol
Formel: C₁₄H₁₇NO₂
MDL: MFCD09878411
CAS-Nummer: 442910-62-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR