2-(3-Chlor-4-fluorphenyl)indol ≥98%

Lieferant: Thermo Scientific

A12920.03 A12920.06
A12920.03EA 56.5 EUR
A12920.03 A12920.06
2-(3-Chlor-4-fluorphenyl)indol ≥98%
2-(3-Chloro-4-fluorophenyl)indole

Purity: 98% CAS No.: 1868-88-8 Molecular Formula: C14H9ClFN Formula Weight: 245.69 Melting Point: 170-173° MDL No.: MFCD00068179

Formel: C₁₄H₉ClFN
Schmelzpunkt: 170…173 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00068179
CAS-Nummer: 1868-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR