2-(3-Chlor-4-fluorphenyl)indol
Lieferant: FLUOROCHEM
008237-1G
008237-5G
FLUO008237-1GEA
224
EUR
FLUO008237-1G
FLUO008237-5G
2-(3-Chlor-4-fluorphenyl)indol
2-(3-Chloro-4-fluorophenyl)indole
Formel:
C₁₄H₉ClFN |
MDL:
MFCD00068179 CAS-Nummer: 1868-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...