1-Boc-6-benzyloxyindol

Lieferant: COMBI-BLOCKS

Synonyme: 6-(Benzyloxy)-1H-indole, N-BOC protected

IN-0164-5G IN-0164-25G IN-0164-1G
COBBIN-0164-5GEA 451 EUR
COBBIN-0164-5G COBBIN-0164-25G COBBIN-0164-1G
1-Boc-6-benzyloxyindol
1-Boc-6-benzyloxyindol
Formel: C₂₀H₂₁NO₃
Molecular Weight: 323,39 g/mol
MDL: MFCD09801032
CAS-Nummer: 933474-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR