2-(4-Fluorphenyl)indol
Lieferant: FLUOROCHEM
008354-1G
008354-5G
FLUO008354-1GEA
69.8
EUR
FLUO008354-1G
FLUO008354-5G
2-(4-Fluorphenyl)indol
2-(4-Fluorophenyl)indole
Formel:
C₁₄H₁₀FN Molecular Weight: 211.24 g/mol |
MDL:
MFCD00068178 CAS-Nummer: 782-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...