Adeninsulfat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Adeninhemisulfat

M-Clarity™ Qualitätsstufe = MQ200

A3159-100G A3159-25G A3159-500G A3159-5G
SIALA3159-100GEA 713 EUR
SIALA3159-100G SIALA3159-25G SIALA3159-500G SIALA3159-5G
Adeninsulfat, Sigma-Aldrich®
Adeninsulfat
Formel: C₅H₅N₅·½H₂SO₄
Molecular Weight: 184,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00213655
CAS-Nummer: 321-30-2
EINECS: 206-286-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR