Adeninsulfat

Lieferant: Serva

Synonyme: Adeninhemisulfat

10741.01 10741.02
SERA10741.01EA 0 EUR
SERA10741.01 SERA10741.02
Adeninsulfat
Adeninsulfat
Formel: C₅H₅N₅·½H₂SO₄
Molecular Weight: 184,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00213655
CAS-Nummer: 321-30-2
EINECS: 206-286-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR