2-Brom-6-(trifluormethoxy)benzothiazol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-6-(trifluoromethoxy)benzo[d]thiazole
HC-2561-5G
HC-2561-1G
HC-2561-250M
COBBHC-2561-5GEA
1580
EUR
COBBHC-2561-5G
COBBHC-2561-1G
COBBHC-2561-250M
2-Brom-6-(trifluormethoxy)benzothiazol
2-Bromo-6-(trifluoromethoxy)benzothiazole
Formel:
C₈H₃BrF₃NOS Molecular Weight: 298,08 g/mol |
MDL:
MFCD09749220 CAS-Nummer: 628725-99-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...