2-Chlor-6-fluorbenzothiazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chloro-6-fluorobenzo[d]thiazole

M-Clarity™ Qualitätsstufe = MQ100

667358-1G 667358-5G
SIAL667358-1GEA 49.7 EUR
SIAL667358-1G SIAL667358-5G
2-Chlor-6-fluorbenzothiazol, Sigma-Aldrich®
2-Chloro-6-fluorobenzothiazole
Formel: C₇H₃ClFNS
Molecular Weight: 187,62 g/mol
MDL: MFCD04448822
CAS-Nummer: 399-74-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR