2-Chlor-6-fluorbenzothiazol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Chloro-6-fluorobenzo[d]thiazole
HC-2280-5G
HC-2280-1G
COBBHC-2280-5GEA
115
EUR
COBBHC-2280-5G
COBBHC-2280-1G
2-Chlor-6-fluorbenzothiazol
2-Chloro-6-fluorobenzothiazole
Formel:
C₇H₃ClFNS Molecular Weight: 187,62 g/mol |
MDL:
MFCD04448822 CAS-Nummer: 399-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...