2-Chlor-6-(trifluormethoxy)benzothiazol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-6-(trifluoromethoxy)benzo[d]thiazole

HC-2560-5G HC-2560-1G
COBBHC-2560-5GEA 391 EUR
COBBHC-2560-5G COBBHC-2560-1G
2-Chlor-6-(trifluormethoxy)benzothiazol
2-Chloro-6-(trifluoromethoxy)benzothiazole
Formel: C₈H₃ClF₃NOS
Molecular Weight: 253.63 g/mol
MDL: MFCD09743918
CAS-Nummer: 133840-96-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR