2-Chlor-6-methoxybenzothiazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chloro-6-methoxy-1,3-benzothiazole, 2-Chloro-6-methoxybenzo[d]thiazole

M-Clarity™ Qualitätsstufe = MQ200

667331-1G 667331-5G
SIAL667331-1GEA 48.6 EUR
SIAL667331-1G SIAL667331-5G
2-Chlor-6-methoxybenzothiazol, Sigma-Aldrich®
2-Chloro-6-methoxybenzothiazole
Formel: C₈H₆ClNOS
Molecular Weight: 199,66 g/mol
MDL: MFCD00671510
CAS-Nummer: 2605-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR