2-Fluorbenzothiazol ≥99%

Lieferant: Thermo Scientific
Gefahr

L19316.03 L19316.06 L19316.MD
L19316.03EA 82.1 EUR
L19316.03 L19316.06 L19316.MD
2-Fluorbenzothiazol ≥99%
2-Fluorobenzothiazole

Purity: 99% CAS No.: 1123-98-4 Molecular Formula: C7H4FNS Formula Weight: 153.18 Refractive Index: 1.5830 Odour: Characteristic Boiling Point: 97-98°/16mm Beilstein Reference: 971503 UN No.: 2810 MDL No.: MFCD00047254

Formel: C₇H₄FNS
Molecular Weight: 153,18 g/mol
Siedepunkt: 97…98 °C (16 mmHg)
Dichte: 1,330
Lagertemperatur: Raumtemperatur
MDL: MFCD00047254
CAS-Nummer: 1123-98-4
EINECS: 214-384-4
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR