1-Ethyl-3-methylimidazoliumtetrachloroaluminat, Sigma-Aldrich®

Lieferant: Merck

51059-1KG-F 51059-100G-F
SIAL51059-1KG-FEA 1010 EUR
SIAL51059-1KG-F SIAL51059-100G-F
1-Ethyl-3-methylimidazoliumtetrachloroaluminat, Sigma-Aldrich®
1-Ethyl-3-methylimidazoliumtetrachloroaluminate
Formel: C₆H₁₁AlCl₄N₂
Molecular Weight: 279.96 g/mol
CAS-Nummer: 80432-05-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR