1-Butyl-3-methylimidazoliumtrifluormethansulfonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Butyl-3-methylimidazolium Trifluoromethanesulphonate
711772-100G
711772-1KG
SIAL711772-100GEA
312
EUR
SIAL711772-100G
SIAL711772-1KG
1-Butyl-3-methylimidazoliumtrifluormethansulfonat, Sigma-Aldrich®
1-Butyl-3-methylimidazoliumtrifluoromethanesulphonate
Formel:
C₉H₁₅F₃N₂O₃S Molecular Weight: 288,29 g/mol |
MDL:
MFCD03427620 CAS-Nummer: 174899-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...