1-Butyl-3-methylimidazoliumtrifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Butyl-3-methylimidazolium Trifluoromethanesulphonate

76420-25G-F 76420-5G-F
SIAL76420-25G-FEA 194 EUR
SIAL76420-25G-F SIAL76420-5G-F
1-Butyl-3-methylimidazoliumtrifluormethansulfonat, Sigma-Aldrich®
1-Butyl-3-methylimidazoliumtrifluoromethanesulphonate
Formel: C₉H₁₅F₃N₂O₃S
Molecular Weight: 288,29 g/mol
MDL: MFCD03427620
CAS-Nummer: 174899-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR