1-Ethyl-3-methylimidazoliumtrifluormethansulfonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Ethyl-3-methylimidazolium trifluoromethanesulphonate
M-Clarity™ Qualitätsstufe = MQ100
04367-250G
04367-50G
04367-5G
SIAL04367-250GEA
1770
EUR
SIAL04367-250G
SIAL04367-50G
SIAL04367-5G
1-Ethyl-3-methylimidazoliumtrifluormethansulfonat, Sigma-Aldrich®
1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate
Formel:
C₇H₁₁F₃N₂O₃S Molecular Weight: 260,24 g/mol |
MDL:
MFCD00210008 CAS-Nummer: 145022-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...