1-Ethyl-3-methylimidazoliumtrifluormethansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Ethyl-3-methylimidazolium trifluoromethanesulphonate

711756-100G 711756-1KG
SIAL711756-100GEA 460 EUR
SIAL711756-100G SIAL711756-1KG
1-Ethyl-3-methylimidazoliumtrifluormethansulfonat, Sigma-Aldrich®
1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate
Formel: C₇H₁₁F₃N₂O₃S
Molecular Weight: 260.24 g/mol
MDL: MFCD00210008
CAS-Nummer: 145022-44-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR