1-Ethyl-3-methylimidazoliumthiocyanat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Ethyl-3-methyl-1H-imidazol-3-ium thiocyanate

43437-1KG-F 43437-100G-F
SIAL43437-1KG-FEA 2630 EUR
SIAL43437-1KG-F SIAL43437-100G-F
1-Ethyl-3-methylimidazoliumthiocyanat, Sigma-Aldrich®
1-Ethyl-3-methylimidazoliumthiocyanate
Formel: C₇H₁₁N₃S
Molecular Weight: 169.25 g/mol
MDL: MFCD06798182
CAS-Nummer: 331717-63-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR