1-Ethyl-3-methylimidazoliumthiocyanat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Ethyl-3-methyl-1H-imidazol-3-ium thiocyanate
M-Clarity™ Qualitätsstufe = MQ100
07424-50G-F
07424-5G-F
SIAL07424-50G-FEA
679
EUR
SIAL07424-50G-F
SIAL07424-5G-F
1-Ethyl-3-methylimidazoliumthiocyanat, Sigma-Aldrich®
1-Ethyl-3-methylimidazoliumthiocyanate
Formel:
C₇H₁₁N₃S Molecular Weight: 169,25 g/mol |
MDL:
MFCD06798182 CAS-Nummer: 331717-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...