2-Brom-4-thiazolcarbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-1,3-thiazole-4-carboxylic acid, 2-Bromothiazole-4-carboxylic acid

HI-1123-5G HI-1123-1G
COBBHI-1123-5GEA 183 EUR
COBBHI-1123-5G COBBHI-1123-1G
2-Brom-4-thiazolcarbonsäure
2-Bromo-4-thiazolecarboxylic acid
Formel: C₄H₂BrNO₂S
Molecular Weight: 208,04 g/mol
MDL: MFCD04115729
CAS-Nummer: 5198-88-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR