2-Brom-5-thiazolcarbonsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Bromothiazole-5-carboxylic acid

H27203.06 H27203.03
H27203.06EA 299 EUR
H27203.06 H27203.03
2-Brom-5-thiazolcarbonsäure ≥97%
2-Bromo-5-thiazolecarboxylic acid

Purity: 97% CAS No.: 54045-76-0 Molecular Formula: C4H2BrNO2S Formula Weight: 208.03 Melting Point: 181-183° MDL No.: MFCD04115730

Formel: C₄H₂BrNO₂S
Molecular Weight: 208,04 g/mol
Schmelzpunkt: 181…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115730
CAS-Nummer: 54045-76-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR