2-Brom-5-thiazolcarbonsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromothiazole-5-carboxylic acid
HI-1124-5G
HI-1124-25G
HI-1124-1G
COBBHI-1124-5GEA
81.1
EUR
COBBHI-1124-5G
COBBHI-1124-25G
COBBHI-1124-1G
2-Brom-5-thiazolcarbonsäure
2-Bromo-5-thiazolecarboxylic acid
Formel:
C₄H₂BrNO₂S Molecular Weight: 208.04 g/mol |
MDL:
MFCD04115730 CAS-Nummer: 54045-76-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...