2-(Trimethylsilyl)thiazol

Lieferant: CHEMPUR SILANE

SILS20279.1 SILS20279.5
SILNSILS20279.1EA 0 EUR
SILNSILS20279.1 SILNSILS20279.5
2-(Trimethylsilyl)thiazol
2-(Trimethylsilyl)thiazole
Formel: C₆H₁₁NSSi
Molecular Weight: 157,31 g/mol
MDL: MFCD00066274
CAS-Nummer: 79265-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR