(S)-(+)-2-Hydroxyisovaleriansäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutanoic acid, (S)-(+)-2-Hydroxy-3-methylbutyric acid

M-Clarity™ Qualitätsstufe = MQ200

379093-1G 379093-5G
SIAL379093-1GEA 71 EUR
SIAL379093-1G SIAL379093-5G
(S)-(+)-2-Hydroxyisovaleriansäure, Sigma-Aldrich®
(S)-(+)-2-Hydroxyisovaleriansäure
Formel: C₅H₁₀O₃
Molecular Weight: 118.13 g/mol
MDL: MFCD00066443
CAS-Nummer: 17407-55-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR