DL-α-Tocopherolacetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme: (±)-α-Tocopherol acetate, all-rac-α-Tocopheryl acetate, Vitamine E acetate, DL-alpha-Tocopheryl acetate, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-yl acetate
M-Clarity™ Qualitätsstufe = MQ200
T3376-100G T3376-5G
SIALT3376-100GEA 241 EUR
SIALT3376-100G SIALT3376-5G
DL-α-Tocopherolacetat, Sigma-Aldrich®
DL-α-Tocopherolacetat
Formel: C₃₁H₅₂O₃
Molecular Weight: 472,75 g/mol
MDL: MFCD00072042
CAS-Nummer: 7695-91-2
EINECS: 231-710-0