2-(2-Hydroxyphenyl)benzothiazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(1,3-Benzothiazol-2-yl)phenol
632589-25G
632589-5G
SIAL632589-25GEA
80.1
EUR
SIAL632589-25G
SIAL632589-5G
2-(2-Hydroxyphenyl)benzothiazol, Sigma-Aldrich®
2-(2-Hydroxyphenyl)benzothiazole
Formel:
C₁₃H₉NOS |
MDL:
MFCD00022869 CAS-Nummer: 3411-95-8 EINECS: 222-299-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...