2-(2-Hydroxyphenyl)benzothiazol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(1,3-Benzothiazol-2-yl)phenol

A19346.06 A19346.14
A19346.06EA 40.2 EUR
A19346.06 A19346.14
2-(2-Hydroxyphenyl)benzothiazol ≥98%
2-(2-Hydroxyphenyl)benzothiazole

Purity: 98% CAS No.: 3411-95-8 Molecular Formula: C13H9NOS Formula Weight: 227.29 Melting Point: 130-134° Beilstein Reference: 173026

Formel: C₁₃H₉NOS
Molecular Weight: 227,29 g/mol
Schmelzpunkt: 130…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022869
CAS-Nummer: 3411-95-8
EINECS: 222-299-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR