β-Jonon

Lieferant: Merck

Synonyme: (E)-4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one, (E)-4-(2,6,6-Trimethylcyclohex-1-en-1-yl)but-3-en-2-one, beta-Ionone

M-Clarity™ Qualitätsstufe = MQ300

W259500-10KG-K W259500-1KG-K W259500-20KG-K
SIALW259500-10KG-KEA 919 EUR
SIALW259500-10KG-K SIALW259500-1KG-K SIALW259500-20KG-K
β-Jonon
β-Jonon
Formel: C₁₃H₂₀O
Molecular Weight: 192,30 g/mol
MDL: MFCD00001549
CAS-Nummer: 79-77-6
EINECS: 201-224-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR