β-Jonon 96%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
(E)-4-(2,6,6-Trimethyl-1-cyclohexenyl)-3-buten-2-one, (E)-4-(2,6,6-Trimethylcyclohex-1-en-1-yl)but-3-en-2-one, beta-Ionone
A13400.22
A13400.36
A13400.22EA
47.5
EUR
A13400.22
A13400.36
β-Jonon 96%
β-Jonon
Purity: 96%
CAS No.: 79-77-6
Molecular Formula: C13H20O
Formula Weight: 192.30
Flash Point: 112°(233°F)
Refractive Index: 1.5200
Melting Point: -49°
Boiling Point: 126-128°/12mm
Beilstein Reference: 1909544
Merck Reference: 14,5056
MDL No.: MFCD00001549
Formel:
C₁₃H₂₀O Molecular Weight: 192,30 g/mol Siedepunkt: 126…128 °C (12 mmHg) Schmelzpunkt: –49 °C Dichte: 0,943 Flammpunkt: 112 °C (233 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001549 CAS-Nummer: 79-77-6 EINECS: 201-224-3 Merck Index: 14,5056 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...