1-Methylimidazoliumhydrogensulfat, Sigma-Aldrich®

Lieferant: Merck

59760-1KG-F 59760-100G-F
SIAL59760-1KG-FEA 1520 EUR
SIAL59760-1KG-F SIAL59760-100G-F
1-Methylimidazoliumhydrogensulfat, Sigma-Aldrich®
1-Methylimidazolium hydrogen sulphate
Formel: C₄H₈N₂O₄S
MDL: MFCD06798198
CAS-Nummer: 681281-87-8
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR