2-Amino-4-phenylthiazol
Lieferant: FLUOROCHEM
Synonyme:
4-Phenylthiazol-2-amine, 4-Phenyl-1,3-thiazol-2-amine
CFF569.10
CFF569.25
CFF569.50
001737-5G
001737-25G
CHMPCFF569.10EA
401
EUR
CHMPCFF569.10
CHMPCFF569.25
CHMPCFF569.50
FLUO001737-5G
FLUO001737-25G
2-Amino-4-phenylthiazol
2-Amino-4-phenylthiazole
Formel:
C₉H₈N₂S Molecular Weight: 176.24 g/mol |
MDL:
MFCD00039680 CAS-Nummer: 2010-06-2 EINECS: 217-926-8 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...