2-Amino-4-phenylthiazol 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Phenylthiazol-2-amine, 4-Phenyl-1,3-thiazol-2-amine
A18488.03
A18488.06
A18488.03EA
35.4
EUR
A18488.03
A18488.06
2-Amino-4-phenylthiazol 98%
2-Amino-4-phenylthiazole
Purity: 98%
CAS No.: 2010-06-2
Molecular Formula: C9H8N2S
Formula Weight: 176.24
Odour: Odourless
Melting Point: 148-152°
Beilstein Reference: 128609
UN No.: 2811
MDL No.: MFCD00039680
Formel:
C₉H₈N₂S Molecular Weight: 176,24 g/mol Schmelzpunkt: 148…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039680 CAS-Nummer: 2010-06-2 EINECS: 217-926-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...