2-Amino-4-phenylthiazol

Lieferant: BIONET RESEARCH

Synonyme: 4-Phenylthiazol-2-amine, 4-Phenyl-1,3-thiazol-2-amine

5K-598S.500 5K-598S.1 5K-598S.5 5K-598S.10
BIOT5K-598S.500EA 0 EUR
BIOT5K-598S.500 BIOT5K-598S.1 BIOT5K-598S.5 BIOT5K-598S.10
2-Amino-4-phenylthiazol
2-Amino-4-phenylthiazole
Formel: C₉H₈N₂S
Molecular Weight: 176,24 g/mol
MDL: MFCD00039680
CAS-Nummer: 2010-06-2
EINECS: 217-926-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR