2-Amino-4-phenylthiazol

Lieferant: Thermo Scientific

Synonyme: 4-Phenylthiazol-2-amine, 4-Phenyl-1,3-thiazol-2-amine

CD00472.1 CD00472.10 CD00472.50
MAYBCD00472.1EA 0 EUR
MAYBCD00472.1 MAYBCD00472.10 MAYBCD00472.50
2-Amino-4-phenylthiazol
2-Amino-4-phenylthiazole
Formel: C₉H₈N₂S
Molecular Weight: 176.24 g/mol
MDL: MFCD00039680
CAS-Nummer: 2010-06-2
EINECS: 217-926-8
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR