5-Brom-2-methylbenzaldehyd

Lieferant: COMBI-BLOCKS

HC-3371-5G HC-3371-1G
COBBHC-3371-5GEA 312 EUR
COBBHC-3371-5G COBBHC-3371-1G
5-Brom-2-methylbenzaldehyd
5-Brom-2-methylbenzaldehyd
Formel: C₈H₇BrO
MDL: MFCD05662390
CAS-Nummer: 90050-59-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR