4-Brom-2-methylbenzaldehyd

Lieferant: COMBI-BLOCKS

CA-4806-1G CA-4806-5G
COBBCA-4806-1GEA 21.9 EUR
COBBCA-4806-1G COBBCA-4806-5G
4-Brom-2-methylbenzaldehyd
4-Bromo-2-methylbenzaldehyde
Formel: C₈H₇BrO
MDL: MFCD07787171
CAS-Nummer: 24078-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR