4-Brom-3-(trifluormethyl)benzonitril

Lieferant: CHEMPUR

05-0068.1 05-0068.5
CHMP05-0068.1EA 0 EUR
CHMP05-0068.1 CHMP05-0068.5
4-Brom-3-(trifluormethyl)benzonitril
4-Brom-3-(trifluormethyl)benzonitril
Formel: C₈H₃BrF₃N
Molecular Weight: 250.02 g/mol
MDL: MFCD03095347
CAS-Nummer: 1735-53-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR