4-Amino-3-(trifluormethyl)benzonitril

Lieferant: FLUOROCHEM

Synonyme: 2-Amino-5-cyanobenzotrifluoride

021886-25G 021886-100G 021886-5G
FLUO021886-25GEA 130 EUR
FLUO021886-25G FLUO021886-100G FLUO021886-5G
4-Amino-3-(trifluormethyl)benzonitril
4-Amino-3-(trifluormethyl)benzonitril
Formel: C₈H₅F₃N₂
Molecular Weight: 186.14 g/mol
MDL: MFCD00275473
CAS-Nummer: 327-74-2
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR