syn-Benzaldehydoxim, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(E)-Benzaldehyde oxime, alpha-Benzaldoxime
M-Clarity™ Qualitätsstufe = MQ200
245674-10G
245674-50G
SIAL245674-10GEA
63.3
EUR
SIAL245674-10G
SIAL245674-50G
syn-Benzaldehydoxim, Sigma-Aldrich®
syn-Benzaldehydoxim
Formel:
C₇H₇NO Molecular Weight: 121,14 g/mol |
MDL:
MFCD00002119 CAS-Nummer: 622-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...