4'-(Trifluormethyl)biphenyl-2-carbonsäure

Lieferant: CHEMPUR

Synonyme: 4-(Trifluoromethyl)-2'-biphenylcarboxylic acid, 4'-(Trifluoromethyl)-2-biphenylcarboxylic acid, 4'-Trifluoromethyl-2-biphenyl carboxylic acid, 4'-(Trifluoromethyl)-[1,1'-biphenyl]-2-carboxylic acid

31-0004.500 31-0004.1
CHMP31-0004.500EA 0 EUR
CHMP31-0004.500 CHMP31-0004.1
4'-(Trifluormethyl)biphenyl-2-carbonsäure
4'-(Trifluormethyl)biphenyl-2-carbonsäure
Formel: C₁₄H₉F₃O₂
Molecular Weight: 266,22 g/mol
MDL: MFCD00075353
CAS-Nummer: 84392-17-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR