2'-(Trifluormethyl)biphenyl-3-carbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2'-(Trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid

CA-4839-5G CA-4839-1G
COBBCA-4839-5GEA 983 EUR
COBBCA-4839-5G COBBCA-4839-1G
2'-(Trifluormethyl)biphenyl-3-carbonsäure
2'-(Trifluormethyl)biphenyl-3-carbonsäure
Formel: C₁₄H₉F₃O₂
Molecular Weight: 266,22 g/mol
MDL: MFCD03424602
CAS-Nummer: 168618-48-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR